man

Test

CAS RN 104451-70-9

FFOF

CAS RN 104451-70-9

IUPAC Name: 2,3,6-trifluorobenzaldehyde
Synonyms: 2,3,6-trifluorobenzaldehyde
Molecular Weight (g/mol): 160.1
Molecular Formula: C7H3F3O
InChi Key: XSBAHBVACIKRTG-UHFFFAOYSA-N
SMILES: FC1=CC=C(F)C(C=O)=C1F
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)