accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10444-89-0

CAS RN 10444-89-0

IUPAC Name: 5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine
Synonyms: 5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol): 169.13
Molecular Formula: C3H2F3N3S
InChi Key: LTEUXHSAYOSFGQ-UHFFFAOYSA-N
SMILES: NC1=NN=C(S1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.