man

Test

CAS RN 10433-06-4

SbH3COH3COH3CO

CAS RN 10433-06-4

IUPAC Name: antimony(3+) triethanolate
Synonyms: antimony(3+) tris(ethoxide)
Molecular Weight (g/mol): 256.94
Molecular Formula: C6H15O3Sb
InChi Key: JGOJQVLHSPGMOC-UHFFFAOYSA-N
SMILES: [Sb+3].CC[O-].CC[O-].CC[O-]
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)