man

Test

CAS RN 10333-68-3

OOHN

CAS RN 10333-68-3

IUPAC Name: 2-(1H-pyrrol-1-yl)benzoic acid
Synonyms: 2-(pyrrol-1-yl)benzoic acid
Molecular Weight (g/mol): 187.2
Molecular Formula: C11H9NO2
InChi Key: GNWTWXOZRSBCOZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1N1C=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results