man

Test

CAS RN 103-23-1

H3CCH3OOOOCH3CH3

CAS RN 103-23-1

IUPAC Name: 1,6-bis(2-ethylhexyl) hexanedioate
Synonyms: BEHA monoplex doa plasthall doa ado (lubricating oil) arlamol doa effomoll DA lankroflex doa flexol plasticizer A-26 hatcol 2908 witamol 320
Molecular Weight (g/mol): 370.57
Molecular Formula: C22H42O4
InChi Key: SAOKZLXYCUGLFA-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC(=O)CCCCC(=O)OCC(CC)CCCC
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)