man

Test

CAS RN 10297-06-0

ClCH

CAS RN 10297-06-0

IUPAC Name: 6-chlorohex-1-yne
Synonyms: 6-chlorohex-1-yne
Molecular Weight (g/mol): 116.59
Molecular Formula: C6H9Cl
InChi Key: ZUKOCGMVJUXIJA-UHFFFAOYSA-N
SMILES: ClCCCCC#C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)