CAS RN 10257-28-0
CAS RN 10257-28-0
IUPAC Name:
6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Synonyms:
d-galactose D-hexose D-hexoses glucose oxidase
Molecular Weight (g/mol):
180.16
Molecular Formula:
C6H12O6
InChi Key:
WQZGKKKJIJFFOK-UHFFFAOYNA-N
SMILES:
OCC1OC(O)C(O)C(O)C1O
1
–
1
of
1
results
1
D-(+)-Galactose Anhydrous, cell culture reagent, 98.5%, MP Biomedicals™
CAS: 10257-28-0 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.16 InChI Key: WQZGKKKJIJFFOK-UHFFFAOYNA-N Synonym: d-galactose,d-galactopyranose,galactose,galactopyranose,galactopyranoside,d-+-galactose,galactose, pure,cerebrose,gal,d-galactopyranoside PubChem CID: 6036 ChEBI: CHEBI:4139