man

Test

CAS RN 10226-30-9

H3COCl

CAS RN 10226-30-9

IUPAC Name: 6-chlorohexan-2-one
Synonyms: 5-chlorovaleroyl
Molecular Weight (g/mol): 134.6
Molecular Formula: C6H11ClO
InChi Key: CMDIDTNMHQUVPE-UHFFFAOYSA-N
SMILES: CC(=O)CCCCCl
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)