CAS RN 102121-60-8
CAS RN 102121-60-8
IUPAC Name:
4-(5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalene-2-amido)benzoic acid
Synonyms:
4-(5,5,8,8-tetramethyl-6,7-dihydronaphthalene-2-amido)benzoic acid
Molecular Weight (g/mol):
351.45
Molecular Formula:
C22H25NO3
InChi Key:
SZWKGOZKRMMLAJ-UHFFFAOYSA-N
SMILES:
CC1(C)CCC(C)(C)C2=CC(=CC=C12)C(=O)NC1=CC=C(C=C1)C(O)=O
1
–
4
of
4
results
2
AM 580, Tocris Bioscience™
CAS: 102121-60-8 Molecular Formula: C22H25NO3 Molecular Weight (g/mol): 351.45 MDL Number: MFCD00673916 InChI Key: SZWKGOZKRMMLAJ-UHFFFAOYSA-N PubChem CID: 2126 ChEBI: CHEBI:64210 IUPAC Name: 4-(5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalene-2-amido)benzoic acid SMILES: CC1(C)CCC(C)(C)C2=CC(=CC=C12)C(=O)NC1=CC=C(C=C1)C(O)=O
3
Tocris Bioscience™ AM 580
AM 580 synthesized to Ancillary Material Grade
4
STEMCELL Technologies AM580
Retinoid pathway activator; Activates retinoic acid receptor (RAR) alpha