man

Test

CAS RN 1014-60-4

CH3H3CCH3H3CH3CCH3

CAS RN 1014-60-4

IUPAC Name: 1,3-di-tert-butylbenzene
Synonyms: 1,3-di-tert-butylbenzene
Molecular Weight (g/mol): 190.33
Molecular Formula: C14H22
InChi Key: ILNDSSCEZZFNGE-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=CC=C1)C(C)(C)C
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)