accessibility menu, dialog, popup

Test

CAS RN 101385-93-7

CAS RN 101385-93-7

IUPAC Name: tert-butyl 3-oxopyrrolidine-1-carboxylate
Synonyms: tert-butyl 3-oxopyrrolidine-1-carboxylate
Molecular Weight (g/mol): 185.22
Molecular Formula: C9H15NO3
InChi Key: JSOMVCDXPUXKIC-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(=O)C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)