CAS RN 10138-93-9
CAS RN 10138-93-9
IUPAC Name:
phosphoric acid; triethylamine
Synonyms:
phosphoric acid; triethylamine
Molecular Weight (g/mol):
199.19
Molecular Formula:
C6H18NO4P
InChi Key:
UNXNGGMLCSMSLH-UHFFFAOYSA-N
SMILES:
OP(O)(O)=O.CCN(CC)CC
1
–
1
of
1
results
1
Honeywell Fluka™ Triethylammonium Phosphate Solution, pH 3, Honeywell™ Fluka™
BioUltra, ∼1.0 M in H2O