accessibility menu, dialog, popup

Test

CAS RN 1008-95-3

NONH2

CAS RN 1008-95-3

IUPAC Name: 4-(1,3-oxazol-5-yl)aniline
Synonyms: 4-(1,3-oxazol-5-yl)aniline
Molecular Weight (g/mol): 160.18
Molecular Formula: C9H8N2O
InChi Key: SLJBMRSOKUTXDF-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C1=CN=CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results