accessibility menu, dialog, popup

Test

CAS RN 1000566-45-9

H3COOHOFFF

CAS RN 1000566-45-9

IUPAC Name: 2-[4-methoxy-3-(trifluoromethyl)phenyl]acetic acid
Synonyms: [4-methoxy-3-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol): 234.17
Molecular Formula: C10H9F3O3
InChi Key: SVJBALZZRCDJHL-UHFFFAOYSA-N
SMILES: COC1=C(C=C(CC(O)=O)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results