Fisher Chemical Formic Acid, 90%, Certified AR for Analysis, Fisher Chemical™
Product Details
318.50 NOK valid until 2025-05-31
Use promo code "24127" to get your promotional price.
Additional Details:
Additional Details: CAS Number: 64-18-6 Weight: 0.61000kg Transport: UN number : 1779 Chem class : 8 Pack group : II
Quantity 500 mL
Product Code. 10775711
Quantity Price
1 440.0  NOK / 500mL
In stock, ready to ship: 2
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Safety and Handling
Description and Specification
Chemical Identifiers
CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number 3297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O
Specification
pH 2.1
CAS 64-18-6
Melting Point 8°C
Boiling Point 101°C
Color Colorless
Molecular Formula CH2O2
MDL Number 3297
Synonym methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt
Synonym methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt
Synonym methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt
Synonym methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt
Synonym methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt
Synonym methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt
Synonym methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt
Synonym methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt
Synonym methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt
Synonym methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
SMILES C(=O)O
IUPAC Name formic acid
Molecular Weight (g/mol) 46.025
PubChem CID 284
ChEBI CHEBI:30751
Formula Weight 46.02g/mol
Quantity 500 mL
Packaging HDPE plastic bottle
Physical Form Liquid
Vapor Pressure 44mbar at 20°C
Alternative Products
Fisherbrand Alternatives
Product Code Brand Description Availability Price
Quantity
10121570 Fisher Chemical Formic acid, 90% 2.5L, Certified AR for analysis 7 1425.00 NOK
Add to basket
Documentation
Technical documents
Safety and Handling
[Formic acid 90%]
[Danger]
[{code=FLAM. LIQ. 3, sequence=1, description=Flammable liquid Category 3}, {code=ACUTE TOX. 4, sequence=2, description=Acute toxicity Category 4}, {code=SKIN CORR. 1A, sequence=3, description=Skin corrosion/irritation Category 1 A}, {code=EYE DAM. 1, sequence=4, description=Serious eye damage/eye irritation Category 1}, {code=ACUTE TOX. 3, sequence=5, description=Acute toxicity Category 3}]
[{code=H226, sequence=1, description=Flammable liquid and vapour.}, {code=H302, sequence=2, description=Harmful if swallowed.}, {code=H314, sequence=3, description=Causes severe skin burns and eye damage.}, {code=H331, sequence=4, description=Toxic if inhaled.}, {code=EUH071, sequence=91, description=Corrosive to the respiratory tract.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=53, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P303+P361+P353, sequence=54, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P305+P351+P338, sequence=55, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=56, description=Immediately call a POISON CENTER/doctor/.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS06, sequence=2, description=SKULL & CROSSBONES}, {code=GHS05, sequence=3, description=CORROSION}]
Product Identifier
  • Formic acid 90%
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 3
Hazard Statement
  • H226- Flammable liquid and vapour.
  • H302- Harmful if swallowed.
  • H314- Causes severe skin burns and eye damage.
  • H331- Toxic if inhaled.
  • EUH071- Corrosive to the respiratory tract.
Precautionary Statement
  • P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310- Immediately call a POISON CENTER/doctor/.