Alternative Packaging
Product Code | Brand | Description | Availability | Price |
| |||
---|---|---|---|---|---|---|---|---|
11345817 | Thermo Scientific Alfa Aesar | ARSENIC (III) OXIDE 99.5% 1KG | 6 | 4115.00 NOK |
Quantity | 250 g |
---|
CAS | 1327-53-3 |
---|---|
Molecular Formula | As2O3 |
Molecular Weight (g/mol) | 197.84 |
MDL Number | MFCD00003433 |
InChI Key | IKWTVSLWAPBBKU-UHFFFAOYSA-N |
Synonym | Arsenic trioxide |
IUPAC Name | diarsorosooxidane |
SMILES | O=[As]O[As]=O |
Boiling Point | 465°C (sublimation) |
---|---|
Chemical Name or Material | Arsenic (III) oxide |
Melting Point | 312.3°C |
CAS | 1327-53-3 |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Notes | (metals basis) |
Molecular Formula | As2O3 |
MDL Number | MFCD00003433 |
UN Number | UN1561 |
Merck Index | 14,804 |
Synonym | Arsenic trioxide |
Solubility Information | Slightly soluble in cold water; Soluble in boiling water,HCl,alkali hydroxide; Insoluble in alcohol,chloroform,and etherSoluble in dilute acids and alkalies; Insoluble in organic solvents; Slightly soluble in water. |
InChI Key | IKWTVSLWAPBBKU-UHFFFAOYSA-N |
SMILES | O=[As]O[As]=O |
IUPAC Name | diarsorosooxidane |
Molecular Weight (g/mol) | 197.84 |
Formula Weight | 197.84 |
Percent Purity | 99.5% |
Odor | Odorless |
Density | 3.74 g/mL |
Product Code | Brand | Description | Availability | Price |
| |||
---|---|---|---|---|---|---|---|---|
11345817 | Thermo Scientific Alfa Aesar | ARSENIC (III) OXIDE 99.5% 1KG | 6 | 4115.00 NOK |