Learn More
Cytiva (formerly Pall Lab) Nylaflo™ Nylon Membrane Disc Filters
A highly versatile laboratory membrane filter.
Brand:  Cytiva (formerly Pall Lab) 66608
Additional Details : Weight : 0.05700kg
 View more versions of this product
Product Code. 17134591
  • 2855.00 NOK / Pack of 100
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
Description
Description
  • Chemical compatibility with esters, bases, and alcohols naturally hydrophilic
  • HPLC certified
  • Provides assurance that the filter will not add artifacts to your analysis
  • For research use only
Specifications
Specifications
Nylon (Hydrophilic)
Non-sterile
114 μm
0.45 μm
100/Pk.
47 mm