Learn More
Fisherbrand™ Glass Vessel for PTFE Tissue Grinder
Use with plunger head for homogenizing tissue material. The glass vessel for PTFE tissue grinder is made from borosilicate glass.
397.38 NOK - 805.00 NOK
Specifications
Quantity | 1 Each |
---|---|
Autoclavable | Yes (individually) |
Color | Clear |
Material | Glass |
Product Type | Tissue Grinder Homogenizer |
Product Code | Brand | Capacity (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
10075911
|
Fisherbrand™
|
2 mL | 120mm |
397.38 NOK
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
10299651
|
Fisherbrand™
|
5 mL | 135mm |
462.94 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
10768992
|
Fisherbrand™
|
15 mL | 155mm |
551.25 NOK
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
||||
10669922
|
Fisherbrand™
|
30 mL | 175mm |
650.00 NOK
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
||||
10331592
|
Fisherbrand™
|
50 mL | 195mm |
805.00 NOK
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
||||
Description
- Precision borosilicate glass
- Ultra pure - no contamination
- Standard clearance 0.15 to 0.25mm
- Volumes stated are working volumes with the plunger in place
Specifications
1 Each | |
Clear | |
Tissue Grinder Homogenizer | |
Tissue Grinder / Homogeniser, PTFE Plain or Serrated Plunger |
Yes (individually) | |
Glass | |
Particle size reduction |